MilliporeSigma
  • A review of the potential protective effects of pentoxifylline against drug-induced nephrotoxicity.
A review of the potential protective effects of pentoxifylline against drug-induced nephrotoxicity.
European journal of clinical pharmacology (2012-11-28)
Zahra Nasiri-Toosi, Simin Dashti-Khavidaki, Hossein Khalili, Mahboob Lessan-Pezeshki
ABSTRACT
Drug-induced nephrotoxicity is one of the most common side effects of medications and accounts for about 20 % of hospital admissions for acute kidney injury (AKI). Some possible pharmacologic mechanisms of pentoxifylline (PTX) that suggest it as a candidate to ameliorate AKI include interaction at the level of the adenosine receptors, increase in erythrocyte deformability, stimulation of vasodilatory prostaglandins production and prevention of vascular congestion, and suppression of tumor necrosis factor alpha (TNF-α) and apoptosis. This manuscript reviews all clinical and animal studies on the use of PTX as a renoprotective agent against a number of nephrotoxic drugs. Data were collected by searching MEDLINE, PubMed, Scopus, Cochrane central register of controlled trials, and Cochrane database systematic reviews. Key words used as search terms were pentoxifylline, nephroprotective, renoprotective, drug-induced renal diseases, drug-induced nephrotoxicity, drug-induced renal toxicity, and drug-induced nephropathy. This search was performed without time limitation. Most greatest number of studies and human clinical trials on the renoprotective effect of PTX against drug-induced nephrotoxicity involves cyclosporine (Cyc). It seems that despite encouraging results from animal studies, there is insufficient evidence to support the renoprotective effect of PTX against Cyc-induced nephrotoxicity in humans. Although some available animal studies show protective effects of PTX against renal toxicity of some antimicrobial and cytotoxic agents, designing clinical trials to approve these nephroprotective effects requires prior confirmation of no reducing antimicrobial or antitumor action of these medications by PTX.
Sigma-Aldrich
Pentoxifylline, solid
USP
Pentoxifylline, United States Pharmacopeia (USP) Reference Standard
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard