MilliporeSigma
  • [In situ photopolymerization of polyacrylamide-based preconcentrator on a microfluidic chip for capillary electrophoresis].
[In situ photopolymerization of polyacrylamide-based preconcentrator on a microfluidic chip for capillary electrophoresis].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (2012-10-02)
Sachio Yamamoto
ABSTRACT
Microchip electrophoresis is widely used for microfluidics and has been studied extensively over the past decade. Translation of capillary electrophoresis methods from traditional capillary systems to a microchip platform provides rapid separation and easy quantitation of sample components. However, most microfluidic systems suffer from critical scaling problems. One promising solution to this problem is online sample preconcentration of all analytes in a sample reservoir before the separation channel. Herein, the following three techniques for online preconcentration during microchip electrophoresis are proposed: (1) in situ fabrication of an ionic polyacrylamide-based preconcentrator on a simple poly(methyl methacrylate) microfluidic chip for perm-selective preconcentration and capillary electrophoretic separation of anionic compounds, (2) simultaneous concentration enrichment and electrophoretic separation of weak acids on a microchip using an in situ photopolymerized carboxylate-type polyacrylamide gels as the perm-selective preconcentrator, and (3) microchip electrophoresis of oligosaccharides using lectin-immobilized preconcentrator gels fabricated by in situ photopolymerization. These techniques are expected to be powerful tools for clinical and pharmaceutical studies with on-line preconcentration during microchip electrophoresis.
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Polyacrylamide, average Mn 40,000
Sigma-Aldrich
Polyacrylamide, average Mn 150,000