MilliporeSigma
Finding the right dose of fulvestrant in breast cancer.
Cancer treatment reviews (2012-07-17)
L Estévez, I Alvarez, I Tusquets, M A Seguí, M Muñoz, Y Fernández, A Lluch
ABSTRACT
Fulvestrant is a selective estrogen receptor downregulator, behaving as a complete antagonist. It was initially approved, at a dose of 250 mg, to treat hormone dependant breast cancer in second line setting. However, a series of pharmacological and pre-clinical studies have suggested that a higher dose of 500 mg may be more effective. The present work summarizes and discusses clinical trials that have aimed to test the benefits of administering fulvestrant at a higher dose. The data support the use of a higher, and more possibly, effective dose of the agent.
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Fulvestrant for system suitability, European Pharmacopoeia (EP) Reference Standard
Fulvestrant, European Pharmacopoeia (EP) Reference Standard