MilliporeSigma
Temozolomide dosing regimens for glioma patients.
Current neurology and neuroscience reports (2012-03-23)
Herwig M Strik, Christine Marosi, Bernd Kaina, Bart Neyns
ABSTRACT
Even in modern times of high-precision brain surgery and irradiation, malignant gliomas belong to the deadliest types of cancer. Due to a marked primary and presumably also acquired resistance, the beneficial effects of cytotoxic chemotherapy are limited. Only one randomized clinical trial demonstrated a significant impact on overall survival with temozolomide. Ever since, there have been attempts to improve the efficacy of alkylating chemotherapy by modulating the distribution of dose in time aiming at a better treatment success. Apart from higher cumulative doses per cycle, better efficacy by depletion of the anti-alkylating O⁶-methylguanine-DNA methyltransferase (MGMT) protein has been a major goal of these regimens. After promising results of single-arm pilot studies, however, randomized studies have been disappointing so far. In this overview, the different strategies of dose-dense temozolomide regimen are highlighted and results of clinical trials put into perspective.
Product Description
Supelco
Temozolomide, VETRANAL®, analytical standard
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)