MilliporeSigma
  • Albendazole associated to surgery or minimally invasive procedures for hydatid disease--how much and how long.
Albendazole associated to surgery or minimally invasive procedures for hydatid disease--how much and how long.
Chirurgia (Bucharest, Romania : 1990) (2012-04-07)
C M Creţu, R R Codreanu, B Mastalier, L G Popa, I Cordoş, M Beuran, D A Steriu Ianulle, S Simion
ABSTRACT
Cystic echinococcosis is parasitic disease due to the larval stage of the metacestode Echinococcus granulosus in the liver (80%), lungs, kidneys, spleen, myocardium etc. Humans are accidental intermediate hosts. Diagnosis is based on laboratory and imaging studies. Treatment is multimodal, surgical and medical. Risks for dangerous complications and relapses are quite common. Adequate follow-up is needed. Albendazole has a central role in the medical therapy of the disease.
Product Description
Albendazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Albendazole, analytical standard, ≥98%
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Albendazole, United States Pharmacopeia (USP) Reference Standard