MilliporeSigma
  • A meta-analysis of topical amphotericin B for the treatment of chronic rhinosinusitis.
A meta-analysis of topical amphotericin B for the treatment of chronic rhinosinusitis.
International forum of allergy & rhinology (2012-01-31)
Seth Isaacs, Samer Fakhri, Amber Luong, Martin J Citardi
ABSTRACT
Fungal species have been implicated in the pathophysiology of chronic rhinosinusitis (CRS). Based on this observation, some clinicians have employed topical antifungal therapy with amphotericin B (AMB); however, its efficacy has not been consistently supported by the literature. A comprehensive review of the English-language literature on the role of topical AMB therapy in CRS treatment was performed. Search criteria included placebo-controlled trials between the years of 2000 and 2009. Studies that reported outcomes based on computed tomography (CT), nasal endoscopy (NE), and symptom scores (SS) were included. Three published studies reported CT outcomes. After pooling patients from these 3 studies, meta-analysis revealed no significant difference between AMB-treated and control groups (p = 0.07). Four studies reported NE outcomes, in which central tendency (mean or median) and variance were identified. Among these 4 studies, combining AMB-treated patients did not reveal any significant effect when compared to the small control group (p = 0.53). Only 2 studies reported SS; no significant differences in symptoms were noted (p = 0.33). A systematic review of the literature does not support the use of topical AMB for the treatment of CRS.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Amphotericin B for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard