MilliporeSigma
Drug safety evaluation of intravitreal triamcinolone acetonide.
Expert opinion on drug safety (2011-11-10)
Daniele Veritti, Andrea Di Giulio, Valentina Sarao, Paolo Lanzetta
ABSTRACT
Triamcinolone acetonide (TA) is a steroidal drug that has been widely administered intravitreally for retinal and choroidal conditions. Safety of steroidal products for intraocular use is essential because of their risk of ocular adverse events. This review comprehensively discusses the safety of intravitreal administration of TA. This paper analyzes the mechanisms of action and key pharmacokinetic attributes and provides a discussion of the main clinical trials investigating clinical applications of intravitreal TA. The safety of intravitreal TA is evaluated through a search of the Medline database from 1980 to 2011. The most relevant literature on the safety of intravitreal TA is also discussed. The complications of intravitreal TA therapy include secondary ocular hypertension in about 20 - 40% of eyes, steroid-induced cataract in about 15 - 20% of cases and postinjection infectious endophthalmitis and pseudoendophthalmitis in less than 1%. TA is an effective drug for various retinal and choroidal diseases when delivered intravitreally. It may imply an off-label use and it may be associated with ocular adverse events. Intravitreal TA is not associated with significant systemic safety risks.
Product Description
Supelco
Triamcinolone acetonide, analytical standard
Triamcinolone acetonide for system suitability, European Pharmacopoeia (EP) Reference Standard
Triamcinolone acetonide, European Pharmacopoeia (EP) Reference Standard
USP
Triamcinolone acetonide, United States Pharmacopeia (USP) Reference Standard