MilliporeSigma
Ezogabine (retigabine).
Nature reviews. Drug discovery (2011-10-01)
Carl E Stafstrom, Seden Grippon, Peter Kirkpatrick
ABSTRACT
Ezogabine (Potiga; Valeant Pharmaceuticals/GlaxoSmithKline), a potassium channel opener, was approved in June 2011 by the U.S. Food and Drug Administration (F.D.A.) for the adjunctive treatment of partial-onset seizures in adult patients. The same drug was granted marketing authorization for this indication in the European Union in March 2011, where it is known as retigabine (Trobalt).
Sigma-Aldrich
Retigabine, ≥98% (HPLC)
Supelco
Retigabine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®