MilliporeSigma
Rosmarinic acid and its derivatives: biotechnology and applications.
Critical reviews in biotechnology (2011-08-16)
Victor P Bulgakov, Yuliya V Inyushkina, Sergey A Fedoreyev
ABSTRACT
Rosmarinic acid (RA) is one of the first secondary metabolites produced in plant cell cultures in extremely high yields, up to 19% of the cell dry weight. More complex derivatives of RA, such as rabdosiin and lithospermic acid B, later were also obtained in cell cultures at high yields. RA and its derivatives possess promising biological activities, such as improvement of cognitive performance, prevention of the development of Alzheimer's disease, cardioprotective effects, reduction of the severity of kidney diseases and cancer chemoprevention. The TNF-α-induced NF-κB signaling pathway has emerged as a central target for RA. Despite these impressive activities and high yields, the biotechnological production of these metabolites on an industrial scale has not progressed. We summarized data suggesting that external stimuli, the Ca(2+)-dependent NADPH oxidase pathway and processes of protein phosphorylation/dephosphorylation are involved in the regulation of biosynthesis of these substances in cultured plant cells. In spite of growing information about pathways regulating biosynthesis of RA and its derivatives in cultured plant cells, the exact mechanism of regulation remains unknown. We suggest that further progress in the biotechnology of RA and its derivatives can be achieved by using new high-throughput techniques.
Product Description
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.