MilliporeSigma
  • Histidine-tag-directed chromophores for tracer analyses in the analytical ultracentrifuge.
Histidine-tag-directed chromophores for tracer analyses in the analytical ultracentrifuge.
Methods (San Diego, Calif.) (2010-12-29)
Lance M Hellman, Chunxia Zhao, Manana Melikishvili, Xiaorong Tao, James E Hopper, Sidney W Whiteheart, Michael G Fried
ABSTRACT
Many recombinant proteins carry an oligohistidine (His(X))-tag that allows their purification by immobilized metal affinity chromatography (IMAC). This tag can be exploited for the site-specific attachment of chromophores and fluorophores, using the same metal ion-nitrilotriacetic acid (NTA) coordination chemistry that forms the basis of popular versions of IMAC. Labeling proteins in this way can allow their detection at wavelengths outside of the absorption envelopes of un-modified proteins and nucleic acids. Here we describe use of this technology in tracer sedimentation experiments that can be performed in a standard analytical ultracentrifuge equipped with absorbance or fluorescence optics. Examples include sedimentation velocity in the presence of low molecular weight chromophoric solutes, sedimentation equilibrium in the presence of high concentrations of background protein and selective labeling to simplify the assignment of species in a complex interacting mixture.
Product Description
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Histidine, European Pharmacopoeia (EP) Reference Standard