MilliporeSigma
  • Fulvestrant: a unique antiendocrine agent for estrogen-sensitive breast cancer.
Fulvestrant: a unique antiendocrine agent for estrogen-sensitive breast cancer.
Expert opinion on pharmacotherapy (2010-02-16)
Peter Kabos, Virginia F Borges
ABSTRACT
The role of estrogen deprivation for the treatment of breast cancer has been understood since the 1800s. Pharmacologic advances in the field in the past decades, including tamoxifen and the aromatase inhibitors, have contributed significantly to the reduced mortality of estrogen-sensitive breast cancer. However, this subtype of breast cancer still presents with relapses and, once metastatic, progression to hormone-refractory state and loss of disease control remain an expected disease course. Fulvestrant, a pure estrogen receptor downregulator, is a new addition to the antiestrogen therapeutic armamentarium since its FDA approval in 2002. Its unique mechanism of action offers potential advantages over other estrogen targeted therapies. Published scientific literature, including presented abstracts, on fulvestrant from 1985 to the present were reviewed with selected publications included. This review addresses current issues and therapies for estrogen-sensitive breast cancer, highlights the role of fulvestrant in current treatment guidelines and outlines some of the ongoing investigations of this compound. Fulvestrant is an effective and well-tolerated drug for treatment of metastatic estrogen-sensitive breast cancer. Work is underway to enhance its clinical benefit to patients as a single agent and in combination with other therapies.
Product Description
Fulvestrant for system suitability, European Pharmacopoeia (EP) Reference Standard
Fulvestrant, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fulvestrant, >98% (HPLC)