MilliporeSigma
Fragrance material review on geraniol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, S P Bhatia, R J Foxenberg, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of geraniol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Geraniol, 98%
Supelco
Geraniol, analytical standard