MilliporeSigma
  • The de novo synthesis of oligosaccharides: application to the medicinal chemistry SAR-study of digitoxin.
The de novo synthesis of oligosaccharides: application to the medicinal chemistry SAR-study of digitoxin.
Current topics in medicinal chemistry (2008-02-22)
Maoquan Zhou, George O'Doherty
ABSTRACT
To address the medicinal chemist's need for new synthetic methods for the preparation of unnatural carbohydrates, a new de novo method for carbohydrate synthesis has been developed. These routes use a palladium catalyzed glycosylation reaction to stereoselectively control the anomeric center and subsequent diastereoselective post glycosylation to install the remaining sugar stereocenters. The utility of this method was demonstrated by the syntheses and biological evaluation of several digitoxin oligosaccharide analogues.
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Digitoxin, European Pharmacopoeia (EP) Reference Standard
USP
Digitoxin, United States Pharmacopeia (USP) Reference Standard