MilliporeSigma
  • A toxicologic and dermatologic assessment of related esters and alcohols of cinnamic acid and cinnamyl alcohol when used as fragrance ingredients.
A toxicologic and dermatologic assessment of related esters and alcohols of cinnamic acid and cinnamyl alcohol when used as fragrance ingredients.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-24)
D Belsito, D Bickers, M Bruze, P Calow, H Greim, J M Hanifin, A E Rogers, J H Saurat, I G Sipes, H Tagami
ABSTRACT
An evaluation and review of a structurally related group of fragrance materials.
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG