MilliporeSigma
Fragrance material review on beta-ionone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
J Lalko, A Lapczynski, D McGinty, S Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of beta-ionone when used as a fragrance ingredient is presented.
Sigma-Aldrich
β-Ionone, 96%
Sigma-Aldrich
β-Ionone, predominantly trans, ≥97%, FCC, FG
Sigma-Aldrich
β-Ionone, natural, ≥95%, FG