MilliporeSigma
Cannabidiol--recent advances.
Chemistry & biodiversity (2007-08-23)
Raphael Mechoulam, Maximilian Peters, Eric Murillo-Rodriguez, Lumír O Hanus
ABSTRACT
The aim of this review is to present some of the recent publications on cannabidiol (CBD; 2), a major non-psychoactive constituent of Cannabis, and to give a general overview. Special emphasis is laid on biochemical and pharmacological advances, and on novel mechanisms recently put forward, to shed light on some of the pharmacological effects that can possibly be rationalized through these mechanisms. The plethora of positive pharmacological effects observed with CBD make this compound a highly attractive therapeutic entity.
Product Description
Supelco
Cannabidiol solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Supelco
Cannabidiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®