MilliporeSigma
Fludarabine phosphate therapy in other lymphoid malignancies.
Seminars in oncology (1990-10-01)
H M Kantarjian, J R Redman, M J Keating
ABSTRACT
The promising results obtained with Fludara I.V. (fludarabine phosphate) treatment in the common indolent B-cell neoplasms have led to their further evaluation in other unusual B-cell malignancies, in Hodgkin's disease, and in T-cell diseases. A significant response rate has been found among patients with macroglobulinemic lymphoma, those with prolymphocytic leukemia or prolymphocytoid variant of chronic lymphocytic leukemia, and those with mycosis fungoides. The limited therapeutic data in hairy-cell leukemia and in Hodgkin's disease are also interesting.
Product Description
Fludarabine for system suitability, European Pharmacopoeia (EP) Reference Standard
Fludarabine phosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fludarabine phosphate