MilliporeSigma
  • Drug Insight: from disturbed motility to disordered movement--a review of the clinical benefits and medicolegal risks of metoclopramide.
Drug Insight: from disturbed motility to disordered movement--a review of the clinical benefits and medicolegal risks of metoclopramide.
Nature clinical practice. Gastroenterology & hepatology (2006-03-03)
P Jay Pasricha, Nonko Pehlivanov, Aravind Sugumar, Joseph Jankovic
ABSTRACT
Metoclopramide, the only drug approved by the FDA for treatment of diabetic gastroparesis, but used off-label for a variety of other gastrointestinal indications, has many potentially troublesome adverse neurologic effects, particularly movement disorders. In this article, we comprehensively review the indications and side effects of metoclopramide, and describe some common pitfalls and strategies to minimize the medicolegal risks to the prescribing physician. Metoclopramide accounts for nearly a third of all drug-induced movement disorders, a common reason for a malpractice suit. The entire spectrum of drug-induced movement disorders, ranging from subtle to life-threatening, can ensue from its use; akathisia and dystonia are generally seen early in the course of metoclopramide-induced movement disorders, whereas tardive dyskinesia and parkinsonism seem to be more prevalent in chronic users. Female sex, age and diabetes are the major risk factors for metoclopramide-induced movement disorders. It is therefore incumbent on gastroenterologists and other prescribing physicians to become familiar with the adverse neurologic effects associated with the use of metoclopramide, and to take appropriate preventive and defensive measures.
Product Description
Supelco
Metoclopramide, VETRANAL®, analytical standard
Metoclopramide, European Pharmacopoeia (EP) Reference Standard