MilliporeSigma
  • Phenobarbital confers its diverse effects by activating the orphan nuclear receptor car.
Phenobarbital confers its diverse effects by activating the orphan nuclear receptor car.
Drug metabolism reviews (2006-05-11)
Susumu Kodama, Masahiko Negishi
ABSTRACT
In the early 1960s, phenobarbital (PB) was shown to induce hepatic drug metabolism and the induction was implicated in the molecular mechanism of drug tolerance development. Since then, it has become evident that PB not only induces drug metabolism, but also triggers pleiotropic effects on liver function, such as cell growth and communication, proliferation of the endoplasmic reticulum, tumor promotion, glucose metabolism, steroid/thyroid hormone metabolism, and bile acid synthesis. Upon activation by PB and numerous PB-type inducers, the nuclear receptor CAR mediates those pleiotropic actions by regulating various hepatic genes, utilizing multiple regulatory mechanisms.
Product Description
Supelco
Phenobarbital solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®