MilliporeSigma
Xylitol, sweeteners, and dental caries.
Pediatric dentistry (2006-05-20)
Kiet A Ly, Peter Milgrom, Marilynn Rothen
ABSTRACT
The purpose of this report was to provide an overview of xylitol and other polyol sweeteners and dental caries for clinicians and to discuss current applications for dental practice and potential community-based public health interventions. Xylitol, like other polyol sweeteners, is a naturally occurring sugar alcohol. Studies suggest polyols are noncariogenic. Furthermore, studies indicate that xylitol can decrease mutans streptococci levels in plaque and saliva and can reduce dental caries in young children, mothers, and in children via their mothers. Food products containing xylitol are now available and have the potential to be widely accessible to consumers to help control rampant decay. Determining whether products contain adequate xylitol amounts for practical use towards prevention is challenging, however, because xylitol content is not clearly labeled. Sufficient evidence exists to support the use of xylitol to reduce caries. Clinicians and dental associations should push for clear recommendations of efficacious dose and frequency of xylitol use and for clear labeling of xylitol content in products to help consumers choose appropriately.
Product Description
Xylitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Xylitol, ≥99% (GC)