MilliporeSigma
  • Risk of surgery after use of potassium chloride for treatment of tubal heterotopic pregnancy.
Risk of surgery after use of potassium chloride for treatment of tubal heterotopic pregnancy.
Obstetrics and gynecology (2006-02-02)
Jerald S Goldstein, Valerie S Ratts, Timothy Philpott, Michael H Dahan
ABSTRACT
Spontaneous heterotopic pregnancies are rare, but with assisted reproductive techniques the incidence may approach 1:100. With the widespread use of transvaginal ultrasonography, physicians have attempted treatment of heterotopic pregnancies with minimally invasive procedures such as transvaginal guided potassium chloride (KCl) injection. However, there are few data on the success of this treatment. A 30-year-old primigravida presented with a desired pregnancy and was found to have a tubal pregnancy in addition to an intrauterine pregnancy. Ultrasound-guided KCl injection into the heterotopic pregnancy was complicated by abdominal pain, surgical abdomen, and hemoperitoneum requiring salpingectomy. A review of the literature revealed that 55% of tubal heterotopic pregnancies treated by KCl injection required subsequent salpingectomy. This raises concerns about the advisability of this treatment.
Product Description
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, ≥99.0%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%