MilliporeSigma
  • Thimerosal-containing vaccines and autistic spectrum disorder: a critical review of published original data.
Thimerosal-containing vaccines and autistic spectrum disorder: a critical review of published original data.
Pediatrics (2004-09-03)
Sarah K Parker, Benjamin Schwartz, James Todd, Larry K Pickering
ABSTRACT
The issue of thimerosal-containing vaccines as a possible cause of autistic spectrum disorders (ASD) and neurodevelopmental disorders (NDDs) has been a controversial topic since 1999. Although most practitioners are familiar with the controversy, many are not familiar with the type or quality of evidence in published articles that have addressed this issue. To assess the quality of evidence assessing a potential association between thimerosal-containing vaccines and autism and evaluate whether that evidence suggests accepting or rejecting the hypothesis, we systematically reviewed published articles that report original data pertinent to the potential association between thimerosal-containing vaccines and ASD/NDDs. Articles for analysis were identified in the National Library of Medicine's Medline database using a PubMed search of the English-language literature for articles published between 1966 and 2004, using keywords thimerosal, thiomersal, mercury, methylmercury, or ethylmercury alone and combined with keywords autistic disorder, autistic spectrum disorder, and neurodevelopment. In addition, we used the "related links" option in PubMed and reviewed the reference sections in the identified articles. All original articles that evaluated an association between thimerosal-containing vaccines and ASD/NDDs or pharmacokinetics of ethylmercury in vaccines were included. Twelve publications that met the selection criteria were identified by the literature search: 10 epidemiologic studies and 2 pharmacokinetic studies of ethylmercury. The design and quality of the studies showed significant variation. The preponderance of epidemiologic evidence does not support an association between thimerosal-containing vaccines and ASD. Epidemiologic studies that support an association are of poor quality and cannot be interpreted. Pharmacokinetic studies suggest that the half-life of ethylmercury is significantly shorter when compared with methylmercury. Studies do not demonstrate a link between thimerosal-containing vaccines and ASD, and the pharmacokinetics of ethylmercury make such an association less likely. Epidemiologic studies that support a link demonstrated significant design flaws that invalidate their conclusions. Evidence does not support a change in the standard of practice with regard to administration of thimerosal-containing vaccines in areas of the world where they are used.
Sigma-Aldrich
Thimerosal, 97.0-101.0% (on dried substance, titration)
Pricing and availability is not currently available.
Thimerosal, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.