MilliporeSigma
Fusidic acid resistance in Staphylococcus aureus.
Archives of disease in childhood (2004-01-08)
D Dobie, J Gray
ABSTRACT
This review summarises current knowledge of the microbiological and clinical aspects of fusidic acid resistance in Staphylococcus aureus, and makes recommendations about fusidic acid prescribing and further research.
Sigma-Aldrich
Fusidic acid
Fusidic acid, European Pharmacopoeia (EP) Reference Standard