MilliporeSigma
Biologically formed amorphous calcium carbonate.
Connective tissue research (2003-09-04)
Steve Weiner, Yael Levi-Kalisman, Sefi Raz, Lia Addadi
ABSTRACT
Many organisms from a wide variety of taxa produce amorphous calcium carbonate (ACC), despite the fact that it is inherently unstable and relatively soluble in its pure state. These properties also make it difficult to detect and characterize ACC. Raman spectroscopy is a particularly useful method for investigating ACC because the sample can be examined wet, and extended X-ray absorption fine structure (EXAFS) analysis can provide detailed information on the short-range order. Other methods for characterizing ACC include infrared spectroscopy, thermogravimetric analysis and differential thermal analysis (TGA and DTA), transmission electron microscopy (TEM), and electron and X-ray diffraction. Because of the difficulties involved, we suspect that ACC is far more widely distributed than is presently known, and a comparison of EXAFS spectra shows that different biogenic ACC phases have different short-range order structures. We also suspect that ACC fulfils many different functions, including as a transient precursor phase during the formation of crystalline calcium carbonate.
Product Description
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®