MilliporeSigma
  • Reactive oxygen species as mediators of organ dysfunction caused by sepsis, acute respiratory distress syndrome, or hemorrhagic shock: potential benefits of resuscitation with Ringer's ethyl pyruvate solution.
Reactive oxygen species as mediators of organ dysfunction caused by sepsis, acute respiratory distress syndrome, or hemorrhagic shock: potential benefits of resuscitation with Ringer's ethyl pyruvate solution.
Current opinion in clinical nutrition and metabolic care (2002-02-15)
Mitchell P Fink
ABSTRACT
Reactive oxygen species are reactive, partly reduced derivatives of molecular oxygen. Important reactive oxygen species in biological systems include superoxide radical anion, hydrogen peroxide, and hydroxyl radical. Peroxynitrite, is another important species in biological systems. A variety of enzymatic and non-enzymatic processes can generate reactive oxygen species in mammalian cells. An extensive body of experimental evidence from studies using animal models supports the view that reactive oxygen species are important in the pathogenesis of ischemia-reperfusion syndromes, sepsis, acute respiratory distress syndrome, and multiple organ dysfunction syndrome. This view is further supported by data from clinical studies that correlate biochemical evidence of reactive oxygen species-mediated stress with the development of acute respiratory distress syndrome or sepsis in patients. Ethyl pyruvate, a simple derivative of pyruvic acid, has been shown to be efficacious in several animal models of critical illness, and warrants further evaluation in this regard.
Sigma-Aldrich
Ethyl pyruvate, natural, FG
Sigma-Aldrich
Ethyl pyruvate, 98%