MilliporeSigma
Ramipril.
International journal of clinical practice (2000-07-27)
W H Smith, S G Ball
ABSTRACT
Ramipril is a long-acting, lipophylic angiotensin converting enzyme inhibitor, its principle action is to inhibit the conversion of angiotensin I to the active angiotensin II. Ramipril is indicated in the treatment of hypertension, congestive cardiac failure (including that following acute myocardial infarction), nephropathy (with and without diabetes mellitus) and now, following the findings of the HOPE study, in the prevention of cardiovascular events (including myocardial infarction) in high risk individuals. This article concentrates on reviewing the evidence supporting ramipril's use in these indications.
Sigma-Aldrich
Ramipril, ≥98% (HPLC)
USP
Ramipril, United States Pharmacopeia (USP) Reference Standard
Ramipril, European Pharmacopoeia (EP) Reference Standard