MilliporeSigma
PPAR-alpha: a key to the mechanism of hepatoprotection by clofibrate.
Toxicological sciences : an official journal of the Society of Toxicology (2000-09-28)
H M Mehendale
ABSTRACT
The article highlighted in this issue is "Peroxisome Proliferator-Activated Receptor Alpha-Null Mice Lack Resistance to Acetaminophen Hepatotoxicity Following Clofibrate Exposure" by Chuan Chen, Gayle E. Hennig, Herbert E. Whiteley, J Christopher Corton, and José E. Manautou (pp. 338-344).
Sigma-Aldrich
Clofibrate, liquid
Supelco
Clofibrate, analytical standard
Clofibrate, European Pharmacopoeia (EP) Reference Standard