MilliporeSigma
  • [Overall and separate clearance--renography with and without captopril].
[Overall and separate clearance--renography with and without captopril].
Revue medicale de Bruxelles (1999-10-19)
M Tondeur
ABSTRACT
The present work briefly describes the radionuclide renal functional tests available for the work up of adult hypertensive patients. The use and specific indications of the different techniques and radiopharmaceuticals are reviewed and summarized. A special attention is given to captopril enhanced renograms: these non invasive procedures contribute to the diagnosis of renovascular hypertension.
Product Description
Captopril for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Captopril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Sigma-Aldrich
Captopril, meets USP testing specifications
Captopril, European Pharmacopoeia (EP) Reference Standard
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material