MilliporeSigma
[Immunomodulation with fumaric acid. Systemic therapy in psoriasis].
Orvosi hetilap (2000-02-29)
B Sebók, I Schneider
ABSTRACT
The treatment of psoriasis vulgaris with fumaric acid esters has been controversial for more than 30 years. Recently the fumaric acid derivatives are marketed antipsoriatics in many European countries. In this paper the clinical efficacy, the side effects as well as the mode of action of these highly potent substances are summarized.
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Sigma-Aldrich
Fumaric acid, FCC, FG
Fumaric acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Fumaric Acid, Pharmaceutical Secondary Standard; Certified Reference Material