- Chlorothalonil: lack of genotoxic potential.
Chlorothalonil: lack of genotoxic potential.
Mutation research (1999-02-25)
H S Rosenkranz
PMID10029695
ABSTRACT
Based upon analyses using a number of validated structure-activity relationship models, it is concluded that the carcinogenicity in rodents of chlorothalonil is not due to a genotoxic mechanism.
MATERIALS