MilliporeSigma
  • Antimicrobial activity of volatile components and various extracts of the red alga Jania rubens.
Antimicrobial activity of volatile components and various extracts of the red alga Jania rubens.
Phytotherapy research : PTR (2006-11-28)
N Ulku Karabay-Yavasoglu, Atakan Sukatar, Guven Ozdemir, Zerrin Horzum
ABSTRACT
The methanol, dichloromethane, hexane, chloroform and volatile oil extracts of the red alga Jania rubens were tested in vitro for their antimicrobial activity (five Gram-positive, four Gram-negative bacteria and Candida albicans ATCC 10239). GC-MS analysis of the volatile components of J. rubens identified 40 compounds which constituted 77.53% of the total. The volatile components of J. rubens consisted of n-docosane (6.35%), n-eicosane (5.77%) and n-tetratriacontane (5.58%) as major components. The methanol and chloroform extracts (4 mg/disc) showed more potent antimicrobial activity than the hexane and dichloromethane extracts and the volatile oil of J. rubens.