MilliporeSigma
  • Characterization of aroma-active compounds in dry flower of Malva sylvestris L. by GC-MS-O analysis and OAV calculations.
Characterization of aroma-active compounds in dry flower of Malva sylvestris L. by GC-MS-O analysis and OAV calculations.
Journal of oleo science (2013-08-30)
Atsushi Usami, Yusei Kashima, Shinsuke Marumoto, Mitsuo Miyazawa
ABSTRACT
In this study, the aroma-active compounds in the dried flower of Malva sylvestris L. were extracted by hydrodistillation and analyzed by gas chromatography-mass spectrometry (GC-MS), and gas chromatography-olfactometry (GC-O) and aroma extraction dilution analysis (AEDA). A light yellow oil with a sweet odor was obtained with a percentage yield of 0.039% (w/w), and 143 volatile compounds (89.86%) were identified by GC-MS. The main compounds were hexadecanoic acid (10.1%), pentacosane (4.8%) and 6,10,14-trimethyl-2-pentadecanone (4.1%). The essential oil consisted mainly of hydrocarbons (25.40%) followed by, alcohols (18.78%), acids (16.66%), ethers (5.01%) ketones (7.28%), esters(12.43%), aldehydes (2.30%) and others (2.00%). Of these compounds, 20 were determined by GC-O and AEDA, to be odor-active (FD (flavor dilution) factor ≥ 1). β-Damascenone (FD = 9, sweet), phenylacetaldehyde (FD = 8, floral, honey-like) and (E)-β-ocimene (FD = 8, spicy) were the most intense aroma-active compounds in M. sylvestris. In order to determine the relative contribution of each of the compounds to the aroma of M. sylvestris, odor activity values (OAVs) were used. β-Damascenone had the highest odor activity values (OAV) (50,700), followed by (E)-β-ionone (15,444) and decanal (3,510). In particular, β-damascenone had a high FD factors, and therefore, this compound was considered to be the main aroma-active components of the essential oil. On the basis of AEDA, OAVs, and sensory evaluation results, β-damascenone is estimated to be the main aroma-active compound of the essential oil.
Sigma-Aldrich
Pentacosane, 99%