MilliporeSigma
  • Novel reversible and selective nerve agent simulant detection in conjunction with superoxide "turn-on" probing.
Novel reversible and selective nerve agent simulant detection in conjunction with superoxide "turn-on" probing.
The Analyst (2014-02-22)
Yoon Jeong Jang, Dhiraj P Murale, David G Churchill
ABSTRACT
Herein, we present fluorescein as a reversible fluorescent sensor for nerve agent simulants diethylchlorophosphate (DCP), diethyl methylphosphonate (DEMP), and diethyl cyanophosphonate (DECP). The superoxide allows for an "off-on" mechanism to regenerate fluorescein. The order of decrease in fluorescence intensity for nerve agent simulants is DCP > DEMP ≫ DECP.