MilliporeSigma
  • Estimation of enantiomeric impurity in piperidin-3-amine by chiral HPLC with precolumn derivatization.
Estimation of enantiomeric impurity in piperidin-3-amine by chiral HPLC with precolumn derivatization.
Chirality (2014-07-22)
Chittabathina Venkata Raghunadha Babu, Nagi Reddy Vuyyuru, K Padmaja Reddy, M V Suryanarayana, K Mukkanti
ABSTRACT
A simple, precise, accurate, robust chiral high-performance liquid chromatographic (chiral HPLC) method was developed for estimation of (S)-piperidin-3-amine (S-isomer) in (R)-piperidin-3-amine dihydrochloride (R-AMP). As AMP is a high-melting solid and nonchromophoric compound, development of a suitable chiral method is a challenging task. The proposed chiral HPLC-UV method involves a precolumn derivatization technique with para toluene sulphonyl chloride (PTSC) in the presence of a base to introduce chromophore into analytes. It utilizes chiralpak AD-H column with a simple mobile phase of 0.1% diethyl amine in ethanol with a 0.5 mL/min flow rate. Analytes were monitored by using a UV detector at 228 nm. The resolution between the two enantiomers was more than 4.0. The developed method was validated as per current International Conference on Harmonization (ICH) guidelines.
Sigma-Aldrich
Sulfuryl chloride, 97%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Sulfuryl chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Diethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%