MilliporeSigma
  • Biocompatible zwitterionic sulfobetaine copolymer-coated mesoporous silica nanoparticles for temperature-responsive drug release.
Biocompatible zwitterionic sulfobetaine copolymer-coated mesoporous silica nanoparticles for temperature-responsive drug release.
Macromolecular rapid communications (2012-04-11)
Jiao-Tong Sun, Zhi-Qiang Yu, Chun-Yan Hong, Cai-Yuan Pan
ABSTRACT
A novel nanocontainer, which could regulate the release of payloads, has been successfully fabricated by attaching zwitterionic sulfobetaine copolymer onto the mesoporous silica nanoparticles (MSNs). RAFT polymerization is employed to prepare the hybrid poly(2-(dimethylamino)ethyl methacrylate)-coated MSNs (MSN-PDMAEMA). Subsequently, the tertiary amine groups in PDMAEMA are quaternized with 1,3-propanesultone to get poly(DMAEMA-co-3-dimethyl(methacryloyloxyethyl)ammonium propanesulfonate)-coated MSNs [MSN-Poly(DMAEMA-co-DMAPS)]. The zwitterionic PDMAPS component endows the nanocarrier with biocompatibility, and the PDMAEMA component makes the copolymer shell temperature-responsive. Controlled release of loaded rhodamine B has been achieved in the saline solutions.
Sigma-Aldrich
1,3-Propanesultone, ≥99%