MilliporeSigma
  • Competition between core-2 GlcNAc-transferase and ST6GalNAc-transferase regulates the synthesis of the leukocyte selectin ligand on human P-selectin glycoprotein ligand-1.
Competition between core-2 GlcNAc-transferase and ST6GalNAc-transferase regulates the synthesis of the leukocyte selectin ligand on human P-selectin glycoprotein ligand-1.
The Journal of biological chemistry (2013-04-04)
Chi Y Lo, Aristotelis Antonopoulos, Rohitesh Gupta, Jun Qu, Anne Dell, Stuart M Haslam, Sriram Neelamegham
ABSTRACT
The binding of selectins to carbohydrate ligands expressed on leukocytes regulates immunity and inflammation. Among the human selectin ligands, the O-linked glycans at the N-terminus of the leukocyte cell-surface molecule P-selectin glycoprotein ligand-1 (PSGL-1, CD162) are important because they bind all selectins (L-, E-, and P-selectin) with high affinity under hydrodynamic shear conditions. Analysis of glycan microheterogeneity at this site is complicated by the presence of 72 additional potential O-linked glycosylation sites on this mucinous protein. To overcome this limitation, truncated forms of PSGL-1, called "PSGL-1 peptide probes," were developed. Ultra-high sensitivity mass spectrometry analysis of glycans released from such probes along with glycoproteomic analysis demonstrate the presence of both the sialyl Lewis-X (sLe(X)) and the di-sialylated T-antigen (NeuAcα2,3Galβ1,3(NeuAcα2,6)GalNAc) at the PSGL-1 N-terminus. Overexpression of glycoprotein-specific ST6GalNAc-transferases (ST6GalNAc1, -2, or -4) in human promyelocytic HL-60 cells altered glycan structures and cell adhesion properties. In particular, ST6GalNAc2 overexpression abrogated cell surface HECA-452/CLA expression, reduced the number of rolling leukocytes on P- and L-selectin-bearing substrates by ~85%, and increased median rolling velocity of remaining cells by 80-150%. Cell rolling on E-selectin was unaltered although the number of adherent cells was reduced by 60%. ST6GalNAc2 partially co-localizes in the Golgi with the core-2 β(1,6)GlcNAc-transferase C2GnT-1. Overall, the data describe the glycan microheterogeneity at the PSGL-1 N-terminus. They suggest that a competition between ST6GalNAc2 and C2GnT-1 for the core-1/Galβ1,3GalNAc glycan may regulate leukocyte adhesion under fluid shear.