MilliporeSigma
  • Evaluation of ¹³C- and ²H-labeled internal standards for the determination of amphetamines in biological samples, by reversed-phase ultra-high performance liquid chromatography-tandem mass spectrometry.
Evaluation of ¹³C- and ²H-labeled internal standards for the determination of amphetamines in biological samples, by reversed-phase ultra-high performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2014-05-02)
Thomas Berg, Morten Karlsen, Ase Marit Leere Oiestad, Jon Eigill Johansen, Huiling Liu, Dag Helge Strand
ABSTRACT
Stable isotope-labeled internal standards (SIL-ISs) are often used when applying liquid chromatography-tandem mass spectrometry (LC-MS/MS) to analyze for legal and illegal drugs. ISs labeled with (13)C, (15)N, and (18)O are expected to behave more closely to their corresponding unlabeled analytes, compared with that of the more classically used (2)H-labeled ISs. This study has investigated the behavior of amphetamine, (2)H3-, (2)H5, (2)H6-, (2)H8-, (2)H11-, and (13)C6-labeled amphetamine, during sample preparation by liquid-liquid extraction and LC-MS/MS analyses. None or only minor differences in liquid-liquid extraction recoveries of amphetamine and the SIL-ISs were observed. The chromatographic resolution between amphetamine and the (2)H-labeled amphetamines increased with the number of (2)H-substitutes. For chromatographic studies we also included seven additional (13)C6-amphetamines and their analytes. All the (13)C6-labeled ISs were co-eluting with their analytes, both when a basic and when an acidic mobile phase were used. MS/MS analyses of amphetamine and its SIL-ISs showed that the ISs with the highest number of (2)H-substitutes required more energy for fragmentation in the collision cell compared with that of the ISs with a lower number. The findings, in this study, support those of previous studies, showing that (13)C-labeled ISs are superior to (2)H-labeled ISs, for analytical purposes.
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Cyclohexane, analytical standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Supelco
(±)-Amphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder