MilliporeSigma
  • Accuracy of contrast-enhanced dual-energy MDCT for the assessment of iodine uptake in renal lesions.
Accuracy of contrast-enhanced dual-energy MDCT for the assessment of iodine uptake in renal lesions.
AJR. American journal of roentgenology (2014-04-25)
Achille Mileto, Daniele Marin, Juan Carlos Ramirez-Giraldo, Emanuele Scribano, Bernhard Krauss, Silvio Mazziotti, Giorgio Ascenti
ABSTRACT
The objective of our study was to assess the accuracy of iodine-related attenuation and iodine quantification as imaging biomarkers of iodine uptake in renal lesions on a single-phase nephrographic image with dual-energy MDCT. Fifty-nine patients (41 men, 18 women; age range, 28-84 years) with 80 renal lesions underwent contrast-enhanced dual-energy CT during the nephrographic phase of enhancement. Renal lesions were characterized as enhancing or nonenhancing on color-coded iodine overlay maps using iodine-related attenuation (in Hounsfield units) and iodine quantification (in milligrams per milliliter). For iodine-related attenuation the iodine uptake thresholds of 15 and 20 HU were tested; a threshold of 0.5 mg/mL was used for iodine quantification. The sensitivity, specificity, positive predictive value (PPV), and negative predictive value (NPV) of iodine-related attenuation and iodine quantification were calculated from chi-square tests of contingency with histopathology or imaging follow-up as the reference standard. The 95% CIs were calculated from binomial expression. Differences in sensitivity and specificity were assessed by means of McNemar analysis. A significant difference in sensitivity and specificity was found between iodine-related attenuation with the thresholds of 15 HU (sensitivity, 91.4%; specificity, 93.3%; PPV, 91.4%; NPV, 93.3%) and 20 HU (sensitivity, 77.1%; specificity, 100%; PPV, 100%; NPV, 84.9%) (p = 0.008) and between iodine quantification (sensitivity, 100%; specificity, 97.7%; PPV, 97.2%; NPV, 100%) and iodine-related attenuation with a threshold of 20 HU (p = 0.004). No significant difference in sensitivity and specificity was found between iodine quantification and iodine-related attenuation with a threshold of 15 HU. Contrast-enhanced dual-energy MDCT with iodine-related attenuation and iodine quantification allows accurate evaluation of iodine uptake in renal lesions on a single-phase nephrographic image.
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm