MilliporeSigma
  • Genome-wide mapping of cellular protein-RNA interactions enabled by chemical crosslinking.
Genome-wide mapping of cellular protein-RNA interactions enabled by chemical crosslinking.
Genomics, proteomics & bioinformatics (2014-04-22)
Xiaoyu Li, Jinghui Song, Chengqi Yi
ABSTRACT
RNA-protein interactions influence many biological processes. Identifying the binding sites of RNA-binding proteins (RBPs) remains one of the most fundamental and important challenges to the studies of such interactions. Capturing RNA and RBPs via chemical crosslinking allows stringent purification procedures that significantly remove the non-specific RNA and protein interactions. Two major types of chemical crosslinking strategies have been developed to date, i.e., UV-enabled crosslinking and enzymatic mechanism-based covalent capture. In this review, we compare such strategies and their current applications, with an emphasis on the technologies themselves rather than the biology that has been revealed. We hope such methods could benefit broader audience and also urge for the development of new methods to study RNA-RBP interactions.
Product Description
Ribonucleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C