MilliporeSigma
  • Agarose gel tailored calcium carbonate nanoparticles-synthesis and biocompatibility evaluation.
Agarose gel tailored calcium carbonate nanoparticles-synthesis and biocompatibility evaluation.
Journal of nanoscience and nanotechnology (2014-04-18)
Santoshkumar Biradar, Virupaxi Goornavar, Adaikkappan Periyakaruppan, Jessica Koehne, Joseph C Hall, Vani Ramesh
ABSTRACT
In this study, a novel approach to tailor the calcium carbonate nanoparticles was exploited based on agarose gel as polymer medium. The size of nanoparticles formed was governed by ionic diffusion and affected by weight percent of agarose and reaction temperature. The size, shape, purity, composition and allotropy of the synthesized nanoparticles were analyzed by different characterization techniques. Purity of nanoparticles as small as 37 nm demonstrates their suitability for broad range of industrial applications. The exposure of rat lung epithelial cells to these nanoparticles even at a higher concentration (50 microg/ml) did not induce considerable oxidative stress or cell death authenticating their fidelity to potential applications in the field of biotechnology and medicine. Through the simple and economic method of synthesis adopted in this study, separation of nanoparticles from the gel was easy, and process parameters could be optimized to control the particle size.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, High EEO, for molecular biology
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.