MilliporeSigma
Fungal phenalenones: chemistry, biology, biosynthesis and phylogeny.
Natural product reports (2014-04-02)
Mahmoud F Elsebai, Muhammad Saleem, Mysore V Tejesvi, Marena Kajula, Sampo Mattila, Mohamed Mehiri, Ari Turpeinen, Anna Maria Pirttilä
ABSTRACT
Covering up to the end of August 2013. Phenalenones are members of a unique class of natural polyketides exhibiting diverse biological potential. This is a comprehensive review of 72 phenalenones with diverse structural features originating from fungal sources. Their bioactive potential and structure elucidation are discussed along with a review of their biosynthetic pathways and the taxonomical relationship between the fungi producing these natural products.
Sigma-Aldrich
Perinaphthenone, 97%