MilliporeSigma
  • Caffeine intake and atrial fibrillation incidence: dose response meta-analysis of prospective cohort studies.
Caffeine intake and atrial fibrillation incidence: dose response meta-analysis of prospective cohort studies.
The Canadian journal of cardiology (2014-04-01)
Min Cheng, Zunsong Hu, Xiangfeng Lu, Jianfeng Huang, Dongfeng Gu
ABSTRACT
The association between habitual caffeine intake with incident atrial fibrillation (AF) was unknown. We conducted a meta-analysis to investigate the association between chronic exposure of caffeine and the risk of AF and to evaluate the potential dose-response relation. We searched PubMed, EMBASE, and the Cochrane Library up to November 2013 and references of relevant retrieved articles. Prospective cohort studies were included with relative risk (RR) or hazard ratio and 95% confidence intervals (CIs) for AF according to coffee/caffeine intake. Six prospective cohort studies with 228,465 participants were included. In the primary meta-analysis, caffeine exposure was weakly associated with a reduced risk of AF (RR, 0.90; 95% CI, 0.81-1.01; P = 0.07; I(2) = 73%). In subgroup analyses, pooled results from studies with adjustment of potential confounders showed an 11% reduction for low doses (RR, 0.89; 95% CI, 0.80-0.99, P = 0.032; I(2) = 30.9%, P = 0.227) and 16% for high doses (RR, 0.84; 95% CI, 0.75-0.94, P = 0.002; I(2) = 24.1%, P = 0.267) of caffeine consumption in AF risk. An inverse relation was found between habitual caffeine intake and AF risk (P for overall trend = 0.015; P for nonlinearity = 0.27) in dose-response meta-analysis and the incidence of AF decreased by 6% (RR, 0.94; 95% CI, 0.90-0.99) for every 300 mg/d increment in habitual caffeine intake. It is unlikely that caffeine consumption causes or contributes to AF. Habitual caffeine consumption might reduce AF risk.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard