MilliporeSigma
  • Field and in vitro insecticidal efficacy of alphacypermethrin-treated high density polyethylene mesh against Culicoides biting midges in South Africa.
Field and in vitro insecticidal efficacy of alphacypermethrin-treated high density polyethylene mesh against Culicoides biting midges in South Africa.
Veterinary parasitology (2014-03-25)
P C Page, K Labuschagne, G J Venter, J P Schoeman, A J Guthrie
ABSTRACT
The efficacy of untreated and alphacypermethrin-treated high density polyethylene (HDPE) mesh against Culicoides biting midges (Diptera: Ceratopogonidae) was determined using Onderstepoort downdraught black light traps and a contact bioassay. Three traps were operated overnight in four replicates of a 3×3 randomised Latin square design near horses under South African field conditions. Both the untreated and alphacypermethrin-treated HDPE mesh significantly (P<0.05) reduced the numbers of Culicoides midges, predominantly Culicoides (Avaritia) imicola Kieffer, collected in the light traps by 4.2 and 7.2 times, respectively. A repellent effect of the alphacypermethrin-treated mesh was not confirmed because the number of midges collected in the light traps with untreated and alphacypermethrin-treated HDPE mesh was not significantly different (P=0.656). Bioassay of the insecticidal contact efficacy indicated median C. imicola mortality of 100% from 30 and 10 min following exposure to the alphacypermethrin-treated HDPE mesh for 1 or 3 min, respectively. In the bioassay, mortality was significantly higher (P=0.016) at 5 min post exposure in the midges exposed to the alphacypermethrin-treated mesh for 3 min (74.8%) compared to the 1 min exposure group (59.5%). The HDPE mesh could be used to reduce exposure of housed animals to Culicoides midges, specifically C. imicola, and viruses transmitted by these midges. Mesh treated with alphacypermethrin had the additional benefit of a rapid insecticidal effect on C. imicola.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Supelco
Cypermethrin solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Polyethylene (LDPE), ERM®, certified reference material
Supelco
Cypermethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cypermethrin, PESTANAL®, analytical standard