MilliporeSigma
  • Genetic markers of toxicity from capecitabine and other fluorouracil-based regimens: investigation in the QUASAR2 study, systematic review, and meta-analysis.
Genetic markers of toxicity from capecitabine and other fluorouracil-based regimens: investigation in the QUASAR2 study, systematic review, and meta-analysis.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-03-05)
Dan Rosmarin, Claire Palles, David Church, Enric Domingo, Angela Jones, Elaine Johnstone, Haitao Wang, Sharon Love, Patrick Julier, Claire Scudder, George Nicholson, Anna Gonzalez-Neira, Miguel Martin, Daniel Sargent, Erin Green, Howard McLeod, Ulrich M Zanger, Matthias Schwab, Michael Braun, Matthew Seymour, Lindsay Thompson, Benjamin Lacas, Valérie Boige, Nuria Ribelles, Shoaib Afzal, Henrik Enghusen, Søren Astrup Jensen, Marie-Christine Etienne-Grimaldi, Gérard Milano, Mia Wadelius, Bengt Glimelius, Hans Garmo, Milena Gusella, Thierry Lecomte, Pierre Laurent-Puig, Eva Martinez-Balibrea, Rohini Sharma, Jesus Garcia-Foncillas, Zdenek Kleibl, Alain Morel, Jean-Pierre Pignon, Rachel Midgley, David Kerr, Ian Tomlinson
ABSTRACT
Fluourouracil (FU) is a mainstay of chemotherapy, although toxicities are common. Genetic biomarkers have been used to predict these adverse events, but their utility is uncertain. We tested candidate polymorphisms identified from a systematic literature search for associations with capecitabine toxicity in 927 patients with colorectal cancer in the Quick and Simple and Reliable trial (QUASAR2). We then performed meta-analysis of QUASAR2 and 16 published studies (n = 4,855 patients) to examine the polymorphisms in various FU monotherapy and combination therapy regimens. Global capecitabine toxicity (grades 0/1/2 v grades 3/4/5) was associated with the rare, functional DPYD alleles 2846T>A and *2A (combined odds ratio, 5.51; P = .0013) and with the common TYMS polymorphisms 5'VNTR2R/3R and 3'UTR 6bp ins-del (combined odds ratio, 1.31; P = 9.4 × 10(-6)). There was weaker evidence that these polymorphisms predict toxicity from bolus and infusional FU monotherapy. No good evidence of association with toxicity was found for the remaining polymorphisms, including several currently included in predictive kits. No polymorphisms were associated with toxicity in combination regimens. A panel of genetic biomarkers for capecitabine monotherapy toxicity would currently comprise only the four DPYD and TYMS variants above. We estimate this test could provide 26% sensitivity, 86% specificity, and 49% positive predictive value-better than most available commercial kits, but suboptimal for clinical use. The test panel might be extended to include additional, rare DPYD variants functionally equivalent to *2A and 2846A, though insufficient evidence supports its use in bolus, infusional, or combination FU. There remains a need to identify further markers of FU toxicity for all regimens.
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Capecitabine, European Pharmacopoeia (EP) Reference Standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Capecitabine, ≥98% (HPLC)
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)