MilliporeSigma
  • Uptake of different crystal structures of TiO₂ nanoparticles by Caco-2 intestinal cells.
Uptake of different crystal structures of TiO₂ nanoparticles by Caco-2 intestinal cells.
Toxicology letters (2014-03-01)
Constantinos Gitrowski, Aliaa R Al-Jubory, Richard D Handy
ABSTRACT
The gastrointestinal uptake of different crystal structures of TiO2 was investigated using Caco-2 intestinal cells. Caco-2 monolayers exhibited time-dependent, saturable uptake of Ti from TiO2 exposures of 1 mgl(-1) over 24h, which was influenced by crystal type. Initial uptake rates were 5.3, 3.73, 3.58 and 4.48 nmol mg(-1)protein h(-1) for bulk, P25, anatase and rutile forms respectively. All exposures caused elevations of Ti in the cells relative to the control (ANOVA P<0.05). Electron micrographs of the Caco-2 monolayer showed the presence of particles inside the cells, and energy dispersive spectroscopy (EDS) confirmed the composition as TiO2. Incubating the cells with 120 IU nystatin (putative endocytosis inhibitor) or 100 μmol l(-1) vanadate (ATPase inhibitor) caused large increases in Ti accumulation for all crystal types relative to controls (ANOVA P<0.05), except for the rutile form with vanadate. Incubating the cells with 90 μmol l(-1) genistein (tyrosine kinase inhibitor) or 27 μmol l(-1) chloropromazine (clathrin-mediated endocytosis inhibitor) caused a large decrease in Ti accumulation relative to the controls (ANOVA P<0.05). Cell viability measures were generally good (low LDH leak, normal cell morphology), but there were some changes in the electrolyte composition (K(+), Na(+), Ca(2+), Mg(2+)) of exposed cells relative to controls. A rise in total Ca(2+) concentration in the cells was observed for all TiO2 crystal type exposures. Overall, the data shows that Ti accumulation for TiO2 NP exposure in Caco-2 cells is crystal structure-dependent, and that the mechanism(s) involves endocytosis of intact particles.
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Supelco
Genistein, analytical standard
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Nystatin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nystatin, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Nystatin, ≥4,400 USP units/mg
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Nystatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
USP
Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
USP
Nystatin, United States Pharmacopeia (USP) Reference Standard
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm