MilliporeSigma
  • Mitomycin C and capecitabine in pretreated patients with metastatic gastric cancer: a multicenter phase II study.
Mitomycin C and capecitabine in pretreated patients with metastatic gastric cancer: a multicenter phase II study.
Journal of cancer research and clinical oncology (2014-02-22)
Manuel Barreto Miranda, Jörg Thomas Hartmann, Salah-Eddin Al-Batran, Melanie Kripp, Deniz Gencer, Andreas Hochhaus, Ralf-Dieter Hofheinz, Kirsten Merx
ABSTRACT
We conducted a multicenter phase II study to assess the toxicity and efficacy of a combination of mitomycin C (MMC) and capecitabine in pretreated patients with metastatic or locally advanced gastric cancer. Thirty-nine patients (77 % male) between 33 and 78 years (median 66) with pretreated locally advanced or metastatic esophagogastric adenocarcinoma and eastern cooperative oncology group performance status of ≤2, measurable lesions, and adequate organ functions were recruited into the study. Eight patients (21 %) had received more than one prior chemotherapy regimen. Treatment consisted of three-weekly MMC 10 mg/m(2) day 1 and capecitabine 2,000 mg/m(2) (day 1-14; repeated day 22). A median of three cycles of therapy was administered. Grade 3 toxicity occurred in 20 patients (54 %). Main grade 3 adverse events were thrombocytopenia (11 %, n = 4), fatigue (8 %, n = 3), and neuropathy (8 %, n = 3). Two events of grade 4 toxicity were reported (5 %) (dyspnea and elevation of alkaline phosphatase due to bone metastases). Partial remission was noticed in 10.3 % (n = 4), stable disease in 33.3 % (n = 13) adding to a tumor control rate of 43.6 %. The median progression-free and overall survival were 2.8 and 5.6 months, respectively. The combination of MMC and capecitabine exhibited a favorable tolerability profile in pretreated patients with gastric cancer. The disease control rate compares adequately with that of other phase II and phase III trials for second-line therapy in gastric cancer. This regimen may be considered as an alternative second-line treatment, especially for patients not suitable for or pretreated with taxanes and/or irinotecan.
Product Description
Mitomycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Capecitabine, ≥98% (HPLC)
Capecitabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture