MilliporeSigma
  • 4-year clinical outcomes and predictors of repeat revascularization in patients treated with new-generation drug-eluting stents: a report from the RESOLUTE All-Comers trial (A Randomized Comparison of a Zotarolimus-Eluting Stent With an Everolimus-Eluting Stent for Percutaneous Coronary Intervention).
4-year clinical outcomes and predictors of repeat revascularization in patients treated with new-generation drug-eluting stents: a report from the RESOLUTE All-Comers trial (A Randomized Comparison of a Zotarolimus-Eluting Stent With an Everolimus-Eluting Stent for Percutaneous Coronary Intervention).
Journal of the American College of Cardiology (2014-02-18)
Masanori Taniwaki, Giulio G Stefanini, Sigmund Silber, Gert Richardt, Pascal Vranckx, Patrick W Serruys, Pawel E Buszman, Henning Kelbaek, Stephan Windecker
ABSTRACT
The aim of the study was to investigate 4-year outcomes and predictors of repeat revascularization in patients treated with the Resolute zotarolimus-eluting stent (R-ZES) (Medtronic, Minneapolis, Minnesota) and XIENCE V everolimus-eluting stent (EES) (Abbott Vascular, Abbott Park, Illinois) in the RESOLUTE (A Randomized Comparison of a Zotarolimus-Eluting Stent With an Everolimus-Eluting Stent for Percutaneous Coronary Intervention) All-Comers trial. Data on long-term outcomes of new-generation drug-eluting stents are limited, and predictors of repeat revascularization due to restenosis and/or progression of disease are largely unknown. Patients were randomly assigned to treatment with the R-ZES (n = 1,140) or the EES (n = 1,152). We assessed pre-specified safety and efficacy outcomes at 4 years including target lesion failure and stent thrombosis. Predictors of revascularization at 4 years were identified by Cox regression analysis. At 4 years, the rates of target lesion failure (15.2% vs. 14.6%, p = 0.68), cardiac death (5.4% vs. 4.7%, p = 0.44), and target vessel myocardial infarction (5.3% vs. 5.4%, p = 1.00), clinically-indicated target lesion revascularization (TLR) (7.0% vs. 6.5%, p = 0.62), and definite/probable stent thrombosis (2.3% vs. 1.6%, p = 0.23) were similar with the R-ZES and EES. Independent predictors of TLR were age, insulin-treated diabetes, SYNTAX (Synergy between PCI with Taxus and Cardiac Surgery) score, treatment of saphenous vein grafts, ostial lesions, and in-stent restenosis. Independent predictors of any revascularization were age, diabetes, previous percutaneous coronary intervention, absence of ST-segment elevation myocardial infarction, smaller reference vessel diameter, SYNTAX score, and treatment of left anterior descending, right coronary artery, saphenous vein grafts, ostial lesions, or in-stent restenosis. R-ZES and EES demonstrated similar safety and efficacy throughout 4 years. TLR represented less than one-half of all repeat revascularization procedures. Patient- and lesion-related factors predicting the risk of TLR and any revascularization showed considerable overlap. (A Randomized Comparison of a Zotarolimus-Eluting Stent With an Everolimus-Eluting Stent for Percutaneous Coronary Intervention [RESOLUTE-AC]; NCT00617084).
Product Description
Supelco
Everolimus solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®