MilliporeSigma
  • Inhibition of biofilm development of uropathogens by curcumin - an anti-quorum sensing agent from Curcuma longa.
Inhibition of biofilm development of uropathogens by curcumin - an anti-quorum sensing agent from Curcuma longa.
Food chemistry (2013-11-23)
Issac Abraham Sybiya Vasantha Packiavathy, Selvam Priya, Shunmugiah Karutha Pandian, Arumugam Veera Ravi
ABSTRACT
Urinary tract infection is caused primarily by the quorum sensing (QS)-dependent biofilm forming ability of uropathogens. In the present investigation, an anti-quorum sensing (anti-QS) agent curcumin from Curcuma longa (turmeric) was shown to inhibit the biofilm formation of uropathogens, such as Escherichia coli, Pseudomonas aeruginosa PAO1, Proteus mirabilis and Serratia marcescens, possibly by interfering with their QS systems. The antibiofilm potential of curcumin on uropathogens as well as its efficacy in disturbing the mature biofilms was examined under light microscope and confocal laser scanning microscope. The treatment with curcumin was also found to attenuate the QS-dependent factors, such as exopolysaccharide production, alginate production, swimming and swarming motility of uropathogens. Furthermore, it was documented that curcumin enhanced the susceptibility of a marker strain and uropathogens to conventional antibiotics.
Product Description
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Curcumin, primary reference standard
Supelco
Curcumin, analytical standard