MilliporeSigma
Patterns of ambulatory care usage and leading treatments for rosacea.
The Journal of dermatological treatment (2013-11-01)
Scott A Davis, Steven R Feldman
ABSTRACT
Millions of rosacea sufferers are not being treated, and the reasons they do not get treatment are not well characterized. The aim of this study is to determine the main reasons for visit, providers seen and treatments used for rosacea. We used data from the National Ambulatory Medical Care Survey for 1993-2010, tabulating the leading reasons for visit, providers seen and treatments used in rosacea visits. There were 1 750 000 visits per year for rosacea. The leading reasons for visit were other diseases of the skin (25.3%), skin rash (19.6%), and discoloration or abnormal pigmentation (14.7%). Dermatologists managed 72.4% of visits. The most common treatments used were topical metronidazole (29.3%), tetracycline (11.0%), minocycline (8.5%), doxycycline (7.9%), and oral metronidazole (6.9%). Some reasons for visit were too nonspecific to provide good insight on why the patient made a visit. Dermatologists manage rosacea most commonly, but primary care physicians need the proper training to diagnose it correctly. Improved strategies to reach untreated people with rosacea are needed.
Product Description
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Doxycycline hyclate
SKU
Pack Size
Availability
Price
Quantity
Supelco
Metronidazole, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Metronidazole, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Metronidazole, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Metronidazole, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Minocycline hydrochloride, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Tetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Doxycycline monohydrate