MilliporeSigma
  • Cemented liner exchange with bone grafting halts the progression of periacetabular osteolysis.
Cemented liner exchange with bone grafting halts the progression of periacetabular osteolysis.
The Journal of arthroplasty (2013-10-01)
Roumen Stamenkov, Susan D Neale, Timothy Kane, David M Findlay, David J Taylor, Donald W Howie
ABSTRACT
The aims of this were to examine the effect of acetabular liner exchange and intra-operative bone grafting surgery on peri-prosthetic osteolysis. Seven patients with well-fixed Harris-Galante-1 acetabular components received cemented exchange liners for worn liners associated with pre-operatively CT-quantified osteolysis. During surgery, accessible osteolytic lesions were debrided and bone-grafted. Except for one patient with recurrent dislocation and acetabular component revision, the other patients had CT scans at a median of 4 months and at approximately 4 years after surgery. None of the pre-operative lesions increased in volume during the post-operative reporting period and no new lesions were detected. These results show that cemented liner exchange surgery can halt the progression of osteolysis and that bone grafting has the potential to restore bone.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Polyethylene (LDPE), ERM®, certified reference material